Gene Transcripts

Gene Symbol
GABRR3
Entrez Gene ID
200959
Gene Name
gamma-aminobutyric acid type A receptor rho3 subunit (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 3: 97986683 - 98035304 on Build GRCh38
UniGene ID
Hs.534578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105580.2 NP_001099050.1 2 195
NR_047685.1 2 195
GenBank mRNA CR749263.1 - 2 204

View Full Product Details