Gene Transcripts

Gene Symbol
C2orf69
Entrez Gene ID
205327
Gene Name
chromosome 2 open reading frame 69
Gene Aliases
Chromosome Location
Chr. 2: 199911256 - 199928273 on Build GRCh38
UniGene ID
Hs.154494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153689.5 NP_710156.3 2 837
GenBank mRNA AK096292.1 - 2 793
BC036456.1 - 2 792 4825616

View Full Product Details