Gene Transcripts

Gene Symbol
OR10Q1
Entrez Gene ID
219960
Gene Name
olfactory receptor family 10 subfamily Q member 1
Gene Aliases
OR11-233
Chromosome Location
Chr. 11: 58227882 - 58228918 on Build GRCh38
UniGene ID
Hs.553646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004471.2 NP_001004471.1 1 542
GenBank mRNA BC140731.1 - 1 542

View Full Product Details