Gene Transcripts

Gene Symbol
PRR15
Entrez Gene ID
222171
Gene Name
proline rich 15
Gene Aliases
Chromosome Location
Chr. 7: 29563796 - 29567300 on Build GRCh38
UniGene ID
Hs.91109
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175887.2 NP_787083.1 2 767
XM_005249653.3 XP_005249710.1 2 1570
XM_011515198.1 XP_011513500.1 2 1565
XM_011515199.1 XP_011513501.1 2 479
XM_017011842.1 XP_016867331.1 2 777
GenBank mRNA BC018144.1 - 2 664
BC029131.1 - 2 767 5189352
FJ857942.1 - 1 87

View Full Product Details