Gene Transcripts

Gene Symbol
SRRM3
Entrez Gene ID
222183
Gene Name
serine/arginine repetitive matrix 3
Gene Aliases
Chromosome Location
Chr. 7: 76201893 - 76287292 on Build GRCh38
UniGene ID
Hs.511025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110199.2 NP_001103669.1 15 2348
NM_001291831.1 NP_001278760.1 16 2427
GenBank mRNA AK092590.1 - 3 766
AK094397.1 - 1 959
BC063788.1 - 1 687
BC096755.1 - 4 877

View Full Product Details