Gene Transcripts

Gene Symbol
IPMK
Entrez Gene ID
253430
Gene Name
inositol polyphosphate multikinase
Gene Aliases
Chromosome Location
Chr. 10: 58191517 - 58269837 on Build GRCh38
UniGene ID
Hs.30280
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152230.4 NP_689416.1 6 1064
XM_011539565.2 XP_011537867.1 6 1105
XM_017016013.1 XP_016871502.1 6 731
GenBank mRNA AF432853.1 - 6 741
AK096839.1 - 3 371
BC065709.1 - 6 1068 6654071

View Full Product Details