Gene Transcripts

Gene Symbol
EHBP1L1
Entrez Gene ID
254102
Gene Name
EH domain binding protein 1 like 1
Gene Aliases
Chromosome Location
Chr. 11: 65576034 - 65592650 on Build GRCh38
UniGene ID
Hs.502867
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099409.1 NP_001092879.1 10 3505
XM_005273867.3 XP_005273924.1 12 1918
XM_005273868.3 XP_005273925.1 11 1743
XM_005273869.3 XP_005273926.1 11 1445
XM_005273870.3 XP_005273927.1 10 1270
XM_006718486.3 XP_006718549.1 11 3542
XM_011544886.2 XP_011543188.1 3 302
XM_017017467.1 XP_016872956.1 11 3292
XM_017017468.1 XP_016872957.1 11 3269
XM_017017469.1 XP_016872958.1 9 1233
GenBank mRNA AK024451.1 - 6 2917
AL713799.1 - 1 2864
AL834433.1 - 10 3202

View Full Product Details