Gene Transcripts

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
Chromosome Location
Chr. 11: 113386014 - 113400418 on Build GRCh38
UniGene ID
Hs.448473
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178510.1 NP_848605.1 8 1814
XM_011542736.2 XP_011541038.1 9 3624
XM_011542737.2 XP_011541039.1 8 3594
XM_011542738.2 XP_011541040.1 9 3882
XM_017017475.1 XP_016872964.1 9 3621
GenBank mRNA AF487542.1 - 8 1720
AJ541797.2 - 8 1814

View Full Product Details