Gene Transcripts

Gene Symbol
OR4C13
Entrez Gene ID
283092
Gene Name
olfactory receptor family 4 subfamily C member 13
Gene Aliases
Chromosome Location
Chr. 11: 49952391 - 49953419 on Build GRCh38
UniGene ID
Hs.553668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001955.2 NP_001001955.2 1 684
GenBank mRNA BC137005.1 - 1 684

View Full Product Details