Gene Transcripts

Gene Symbol
FAM102B
Entrez Gene ID
284611
Gene Name
family with sequence similarity 102 member B
Gene Aliases
SYM-3B
Chromosome Location
Chr. 1: 108560349 - 108639327 on Build GRCh38
UniGene ID
Hs.200230
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010883.2 NP_001010883.2 2, 3 330
GenBank mRNA BC127898.1 - 2, 3 330
BC140863.1 - 2, 3 186 9021731
BC140865.1 - 2, 3 186 9021733
BC150530.1 - 2, 3 330 40132919

View Full Product Details