Gene Transcripts

Gene Symbol
RUSC1-AS1
Entrez Gene ID
284618
Gene Name
RUSC1 antisense RNA 1
Gene Aliases
C1orf104
Chromosome Location
Chr. 1: 155320460 - 155324147 on Build GRCh38
UniGene ID
Hs.650801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039517.1 NP_001034606.1 2 687
GenBank mRNA BC062571.1 - 2 716
BC080538.1 - 2 687 5018492

View Full Product Details