Gene Transcripts

Gene Symbol
IGSF10
Entrez Gene ID
285313
Gene Name
immunoglobulin superfamily member 10
Gene Aliases
CMF608
Chromosome Location
Chr. 3: 151435051 - 151619926 on Build GRCh38
UniGene ID
Hs.58561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178145.1 NP_001171616.1 2 880
NM_001178146.1 NP_001171617.1 2 727
NM_178822.4 NP_849144.2 6 6627
XM_011512708.2 XP_011511010.1 12 7436
XM_011512709.2 XP_011511011.1 11 7167
GenBank mRNA AK093069.1 - 2 863
AK098838.1 - 1 70
AK128160.1 - 3 2676
AY273815.1 - 6 6627
BC031063.1 - 2 717
BC045734.1 - 2 718

View Full Product Details