Gene Transcripts

Gene Symbol
TCAIM
Entrez Gene ID
285343
Gene Name
T-cell activation inhibitor, mitochondrial
Gene Aliases
C3orf23, TOAG-1, TOAG1
Chromosome Location
Chr. 3: 44338119 - 44409448 on Build GRCh38
UniGene ID
Hs.55131
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029839.2 NP_001025010.1 3 569
NM_001029840.2 NP_001025011.1 3 387
NM_001282913.1 NP_001269842.1 3 387
NM_001282914.1 NP_001269843.1 3 569
NM_001282915.1 NP_001269844.1 3 387
NM_173826.3 NP_776187.2 3 569
XM_017006251.1 XP_016861740.1 3 614
GenBank mRNA AK056593.1 - 3 569
AK292756.1 - 3 387
AL832473.1 - 3 246
AL833118.1 - 1 59
BC009400.2 - 3 365
BC038420.1 - 3 489
BC047231.1 - 3 264 4817904
BC062587.1 - 3 541
BC073829.1 - 3 214
BC101058.2 - 3 246
BC101059.2 - 3 246 40008143
BC101060.2 - 3 246 40008144
BC101061.2 - 3 246 40008152
CN372173.1 - 3 342
DC413255.1 - 3 387

View Full Product Details