Gene Transcripts
- Gene Symbol
- LSMEM1
- Entrez Gene ID
- 286006
- Gene Name
- leucine rich single-pass membrane protein 1
- Gene Aliases
- C7orf53
- Chromosome Location
- Chr. 7: 112480219 - 112491062 on Build GRCh38
- UniGene ID
- Hs.396189
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134468.1 |
NP_001127940.1
|
3, 4 |
385 |
|
NM_182597.2 |
NP_872403.1
|
3, 4 |
591 |
|
XM_011516074.1 |
XP_011514376.1
|
3, 4 |
1094 |
|
XM_011516075.2 |
XP_011514377.1
|
3, 4 |
2781 |
|
XM_011516076.2 |
XP_011514378.1
|
3, 4 |
2417 |
|
XM_017012028.1 |
XP_016867517.1
|
2, 3 |
2299 |
|
GenBank mRNA |
AK096894.1 |
- |
3, 4 |
607 |
|
BC031976.1 |
- |
5, 6 |
658 |
|
BC126149.1 |
- |
3, 4 |
429 |
8991865 |
BC130568.1 |
- |
3, 4 |
429 |
40146599 |
DB456503.2 |
- |
3, 4 |
385 |
|
View Full Product Details