Gene Transcripts

Gene Symbol
VN1R5
Entrez Gene ID
317705
Gene Name
vomeronasal 1 receptor 5 (gene/pseudogene)
Gene Aliases
V1RL5
Chromosome Location
Chr. 1: 247256072 - 247257145 on Build GRCh38
UniGene ID
Hs.553686
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173858.1 NP_776257.1 1 82
GenBank mRNA BC126341.1 - 1 182
BC126367.1 - 1 182 8992083

View Full Product Details