Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143679.1 | NP_001137151.1 | 5 | 693 | |
NM_001143680.1 | NP_001137152.1 | 5 | 489 | ||
NM_001143681.1 | NP_001137153.1 | 5 | 396 | ||
NM_015917.2 | NP_057001.1 | 6 | 525 | ||
GenBank mRNA | AF068287.1 | - | 6 | 512 | |
AF070657.1 | - | 6 | 483 | ||
AF087849.1 | - | 6 | 497 | ||
AK000640.1 | - | 6 | 483 | ||
AK095263.1 | - | 5 | 782 | ||
AK289570.1 | - | 5 | 626 | ||
AK289580.1 | - | 6 | 463 | ||
AK295592.1 | - | 5 | 384 | ||
AK299968.1 | - | 5 | 407 | ||
AL136938.1 | - | 6 | 511 | ||
AY520571.1 | - | 6 | 494 | ||
BC001231.1 | - | 6 | 472 | 3460150 | |
BC050715.1 | - | 6 | 445 | 6061265 | |
BC063425.1 | - | 5 | 622 | 5218251 | |
BC110600.1 | - | 4 | 281 | ||
BQ436828.1 | - | 5 | 489 | ||
CR533445.1 | - | 6 | 440 |