Gene Transcripts

Gene Symbol
SPEM1
Entrez Gene ID
374768
Gene Name
spermatid maturation 1
Gene Aliases
C17orf83
Chromosome Location
Chr. 17: 7420360 - 7421632 on Build GRCh38
UniGene ID
Hs.710599
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199339.2 NP_955371.2 1 146
GenBank mRNA AK097400.1 - 1 182
BC033882.1 - 1 146 5742860

View Full Product Details