Gene Transcripts
- Gene Symbol
- FAM21A
- Entrez Gene ID
- 387680
- Gene Name
- family with sequence similarity 21 member A
- Gene Aliases
- FAM21B, bA56A21.1, bA98I6.1
- Chromosome Location
- Chr. 10: 50067888 - 50133509 on Build GRCh38
- UniGene ID
- Hs.365286
- Species
- Homo sapiens
- Gene Symbol
- FAM21C
- Entrez Gene ID
- 253725
- Gene Name
- family with sequence similarity 21 member C
- Gene Aliases
- FAM21A, VPEF
- Chromosome Location
- Chr. 10: 45727170 - 45792964 on Build GRCh38
- UniGene ID
- Hs.365286
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC150611.1 |
- |
3 |
290 |
|
View Full Product Details