Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011720.1 | NP_001011720.1 | 5 | 1333 | |
NM_001287258.1 | NP_001274187.1 | 6 | 1451 | ||
NM_001287259.1 | NP_001274188.1 | 6 | 1443 | ||
NM_001287260.1 | NP_001274189.1 | 5 | 1187 | ||
XM_006716447.3 | XP_006716510.1 | 3 | 683 | ||
XM_011517524.2 | XP_011515826.1 | 5 | 1316 | ||
XM_011517525.2 | XP_011515827.1 | 5 | 1178 | ||
XM_017013404.1 | XP_016868893.1 | 4 | 1550 | ||
GenBank mRNA | AY534247.1 | - | 5 | 1333 | |
BC137104.1 | - | 5 | 1251 | 9021101 | |
BC137105.1 | - | 5 | 1187 | 9021102 | |
BC151845.1 | - | 6 | 1410 |