Gene Transcripts

Gene Symbol
OR5D13
Entrez Gene ID
390142
Gene Name
olfactory receptor family 5 subfamily D member 13 (gene/pseudogene)
Gene Aliases
Chromosome Location
Chr. 11: 55773438 - 55774382 on Build GRCh38
UniGene ID
Hs.553743
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001967.1 NP_001001967.1 1 235

View Full Product Details