Gene Transcripts

Gene Symbol
OR5AU1
Entrez Gene ID
390445
Gene Name
olfactory receptor family 5 subfamily AU member 1
Gene Aliases
OR14-38
Chromosome Location
Chr. 14: 21154937 - 21156025 on Build GRCh38
UniGene ID
Hs.553768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004731.1 NP_001004731.1 1 615
GenBank mRNA BC137304.1 - 1 653
BC137305.1 - 1 653 9021302

View Full Product Details