Gene Transcripts

Gene Symbol
GLOD5
Entrez Gene ID
392465
Gene Name
glyoxalase domain containing 5
Gene Aliases
Chromosome Location
Chr. X: 48761572 - 48773648 on Build GRCh38
UniGene ID
Hs.437539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080489.2 NP_001073958.2 4 459
XM_011543911.2 XP_011542213.1 5 562
XM_011543912.2 XP_011542214.1 5 706

View Full Product Details