Gene Transcripts

Gene Symbol
VWC2L
Entrez Gene ID
402117
Gene Name
von Willebrand factor C domain containing protein 2-like
Gene Aliases
Chromosome Location
Chr. 2: 214411737 - 214575929 on Build GRCh38
UniGene ID
Hs.534834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080500.2 NP_001073969.1 3 615
GenBank mRNA AB374231.1 - 2 485
BC146903.1 - 3 615 9056831
BC146911.1 - 3 615 9056839
BC146931.1 - 3 615 9056859
BC146935.1 - 3 615 9056863
EU541473.1 - 2 488

View Full Product Details