Gene Transcripts

Gene Symbol
OR5H15
Entrez Gene ID
403274
Gene Name
olfactory receptor family 5 subfamily H member 15
Gene Aliases
Chromosome Location
Chr. 3: 98168700 - 98169641 on Build GRCh38
UniGene ID
Hs.554713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005515.1 NP_001005515.1 1 698

View Full Product Details