Gene Transcripts

Gene Symbol
C10orf62
Entrez Gene ID
414157
Gene Name
chromosome 10 open reading frame 62
Gene Aliases
bA548K23.1
Chromosome Location
Chr. 10: 97589693 - 97590934 on Build GRCh38
UniGene ID
Hs.662302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009997.2 NP_001009997.2 1 422
GenBank mRNA BC037252.1 - 1 388
BC043281.1 - 1 873
BI826342.1 - 1 421

View Full Product Details