Gene Transcripts

Gene Symbol
OR11H12
Entrez Gene ID
440153
Gene Name
olfactory receptor family 11 subfamily H member 12
Gene Aliases
Chromosome Location
Chr. 14: 18601117 - 18602097 on Build GRCh38
UniGene ID
Hs.534880
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013354.1 NP_001013372.1 1 654
GenBank mRNA BC136730.1 - 1 726
BC136731.1 - 1 726 9020721

Gene Symbol
OR11H1
Entrez Gene ID
81061
Gene Name
olfactory receptor family 11 subfamily H member 1
Gene Aliases
OR11H12, OR22-1
Chromosome Location
Chr. 22: 15528159 - 15529139 on Build GRCh38
UniGene ID
Hs.554706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005239.1 NP_001005239.1 1 654

Gene Symbol
OR11H2
Entrez Gene ID
79334
Gene Name
olfactory receptor family 11 subfamily H member 2
Gene Aliases
C14orf15, OR11H13, OR11H2P, OR11H8P
Chromosome Location
Chr. 14: 19712904 - 19714332 on Build GRCh38
UniGene ID
Hs.554706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197287.1 NP_001184216.1 2 726

View Full Product Details