Gene Transcripts
- Gene Symbol
- OR4Q3
- Entrez Gene ID
- 441669
- Gene Name
- olfactory receptor family 4 subfamily Q member 3
- Gene Aliases
- C14orf13, HSA6, OR14-3, OR4Q4, c14_5008
- Chromosome Location
- Chr. 14: 19747428 - 19748369 on Build GRCh38
- UniGene ID
- Hs.553828
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172194.1 |
NP_751944.1
|
1 |
339 |
|
View Full Product Details