Gene Transcripts
- Gene Symbol
- GAS1
- Entrez Gene ID
- 2619
- Gene Name
- growth arrest specific 1
- Gene Aliases
- Chromosome Location
- Chr. 9: 86944362 - 86947189 on Build GRCh38
- UniGene ID
- Hs.65029
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002048.2 |
NP_002039.2
|
1 |
1994 |
|
GenBank mRNA |
L13698.1 |
- |
1 |
1994 |
|
View Full Product Details