Gene Transcripts

Gene Symbol
SERPINE3
Entrez Gene ID
647174
Gene Name
serpin family E member 3
Gene Aliases
Chromosome Location
Chr. 13: 51334976 - 51362391 on Build GRCh38
UniGene ID
Hs.685891
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101320.1 NP_001094790.1 2 331
XM_017020707.1 XP_016876196.1 1 9291
GenBank mRNA AM402969.1 - 2 274
AX772926.1 - 2 331

View Full Product Details