Gene Transcripts
- Gene Symbol
- MROH3P
- Entrez Gene ID
- 647215
- Gene Name
- maestro heat like repeat family member 3, pseudogene
- Gene Aliases
- C1orf81
- Chromosome Location
- Chr. 1: 200917064 - 200966676 on Build GRCh38
- UniGene ID
- Hs.576504
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ983818.1 |
- |
15 |
1964 |
|
View Full Product Details