Gene Transcripts

Gene Symbol
GPR89B
Entrez Gene ID
51463
Gene Name
G protein-coupled receptor 89B
Gene Aliases
GPHR, GPR89, GPR89C, SH120, UNQ192
Chromosome Location
Chr. 1: 147928393 - 147993523 on Build GRCh38
UniGene ID
Hs.504320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016334.3 NP_057418.1 4 363
XM_005277401.3 XP_005277458.1 4 336
XM_005277402.4 XP_005277459.1 4 500
XM_005277404.3 XP_005277461.1 4 336
XM_006711377.3 XP_006711440.1 5 399
XM_006711378.2 XP_006711441.1 5 382
XM_011509610.2 XP_011507912.1 5 396
XM_011509612.2 XP_011507914.1 5 337
XM_011509613.2 XP_011507915.1 4 336
XM_017001448.1 XP_016856937.1 5 899
XM_017001449.1 XP_016856938.1 4 336
GenBank mRNA AK299995.1 - 4 334
AK302266.1 - 4 355
AL133020.1 - 4 347
DC305701.1 - 4 363
KJ534849.1 - 4 219
KJ534850.1 - 4 219
KJ534851.1 - 4 219
KJ535070.1 - 5 547
U78723.1 - 4 322

Gene Symbol
GPR89A
Entrez Gene ID
653519
Gene Name
G protein-coupled receptor 89A
Gene Aliases
GPHR, GPR89, GPR89B, SH120, UNQ192
Chromosome Location
Chr. 1: 145607960 - 145670977 on Build GRCh38
UniGene ID
Hs.504320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097612.1 NP_001091081.1 4 393
NM_001097613.2 NP_001091082.2 5 439
NR_036541.1 4 393
XM_006711492.3 XP_006711555.1 4 363
XM_006711493.3 XP_006711556.1 5 351
XM_011509908.2 XP_011508210.1 5 409
XM_011509909.2 XP_011508211.1 4 621
XM_011509910.2 XP_011508212.1 4 363
XM_017002150.1 XP_016857639.1 4 363
GenBank mRNA AB097024.1 - 4 334
AB097025.1 - 4 334
AF132947.1 - 4 318
AK021758.1 - 4 363
AK223223.1 - 4 334
AK297145.1 - 4 393
AK301939.1 - 4 240
AK316371.1 - 5 351
AY358966.1 - 4 368
BC003187.1 - 4 296 3537949
BC067816.1 - 4 332 5295014
BC146880.1 - 4 314 9056808
BT006679.1 - 4 219
DC349534.1 - 4 393
KJ534847.1 - 4 219
KJ534848.1 - 4 219
KJ535029.1 - 4 219
KJ535049.1 - 4 219

View Full Product Details