Gene Transcripts
- Gene Symbol
- ARAF
- Entrez Gene ID
- 369
- Gene Name
- A-Raf proto-oncogene, serine/threonine kinase
- Gene Aliases
- A-RAF, ARAF1, PKS2, RAFA1
- Chromosome Location
- Chr. X: 47561100 - 47571921 on Build GRCh38
- UniGene ID
- Hs.446641
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256196.1 |
NP_001243125.1
|
4, 5 |
512 |
|
NM_001256197.1 |
NP_001243126.1
|
4, 5 |
512 |
|
NM_001654.4 |
NP_001645.1
|
4, 5 |
512 |
|
XM_006724529.2 |
XP_006724592.1
|
4, 5 |
433 |
|
XM_011543906.2 |
XP_011542208.1
|
4, 5 |
1823 |
|
XM_011543908.2 |
XP_011542210.1
|
4, 5 |
1822 |
|
GenBank mRNA |
AB158254.1 |
- |
3, 4 |
309 |
|
AB208831.1 |
- |
4, 5 |
409 |
|
AK130043.1 |
- |
4, 5 |
405 |
|
AK297453.1 |
- |
4, 5 |
405 |
|
AK300975.1 |
- |
4, 5 |
409 |
|
AK307747.1 |
- |
4, 5 |
405 |
|
BC002466.2 |
- |
4, 5 |
392 |
3347509 |
BC007514.2 |
- |
4, 5 |
390 |
2966604 |
BT019864.1 |
- |
3, 4 |
301 |
|
DA264472.1 |
- |
4, 5 |
434 |
|
DC362427.1 |
- |
4, 5 |
512 |
|
KU177914.1 |
- |
3, 4 |
301 |
|
KU177915.1 |
- |
3, 4 |
301 |
|
X04790.1 |
- |
4, 5 |
495 |
|
View Full Product Details