Gene Transcripts
- Gene Symbol
- CT45A8
- Entrez Gene ID
- 102723737
- Gene Name
- cancer/testis antigen family 45, member A8
- Gene Aliases
- Chromosome Location
- Chr. X: 135846497 - 135854588 on Build GRCh38
- UniGene ID
- Hs.568346
- Species
- Homo sapiens
- Gene Symbol
- CT45A10
- Entrez Gene ID
- 102723631
- Gene Name
- cancer/testis antigen family 45, member A10
- Gene Aliases
- Chromosome Location
- Chr. X: 135880752 - 135893577 on Build GRCh38
- UniGene ID
- Hs.535081
- Species
- Homo sapiens
- Gene Symbol
- CT45A2
- Entrez Gene ID
- 728911
- Gene Name
- cancer/testis antigen family 45, member A2
- Gene Aliases
- CT45-2, CT45.2
- Chromosome Location
- Chr. X: 135811979 - 135820062 on Build GRCh38
- UniGene ID
- Hs.535081
- Species
- Homo sapiens
- Gene Symbol
- CT45A3
- Entrez Gene ID
- 441519
- Gene Name
- cancer/testis antigen family 45, member A3
- Gene Aliases
- CT45-3, CT45-4, CT45.3, CT45.4, CT45A4
- Chromosome Location
- Chr. X: 135760067 - 135768222 on Build GRCh38
- UniGene ID
- Hs.535080
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098689.1 |
- |
3 |
458 |
|
- Gene Symbol
- CT45A6
- Entrez Gene ID
- 541465
- Gene Name
- cancer/testis antigen family 45, member A6
- Gene Aliases
- CT45-6, CT45.6
- Chromosome Location
- Chr. X: 135794687 - 135802755 on Build GRCh38
- UniGene ID
- Hs.676560
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY743714.1 |
- |
3 |
613 |
|
- Gene Symbol
- CT45A9
- Entrez Gene ID
- 102723680
- Gene Name
- cancer/testis antigen family 45, member A9
- Gene Aliases
- Chromosome Location
- Chr. X: 135863418 - 135871812 on Build GRCh38
- UniGene ID
- Hs.632281
- Species
- Homo sapiens
View Full Product Details