Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_011511771.2 | XP_011510073.1 | 4 | 814 | |
XM_011511772.1 | XP_011510074.1 | 6 | 488 | ||
XM_011511773.1 | XP_011510075.1 | 2 | 159 | ||
XM_011511776.1 | XP_011510078.1 | 5 | 423 | ||
XM_017004851.1 | XP_016860340.1 | 6 | 498 | ||
XM_017004852.1 | XP_016860341.1 | 3 | 285 | ||
XM_017004853.1 | XP_016860342.1 | 6 | 498 | ||
XM_017004854.1 | XP_016860343.1 | 6 | 498 |