Gene Transcripts

Gene Symbol
KRT18P55
Entrez Gene ID
284085
Gene Name
keratin 18 pseudogene 55
Gene Aliases
Chromosome Location
Chr. 17: 28275986 - 28307382 on Build GRCh38
UniGene ID
Hs.371796
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028334.1 2 427
GenBank mRNA AK097823.1 - 2 428
BC112026.1 - 2 48 8327494
BC112028.1 - 2 36 8327496

View Full Product Details