Gene Transcripts

Gene Symbol
OR4F16
Entrez Gene ID
81399
Gene Name
olfactory receptor family 4 subfamily F member 16
Gene Aliases
OR1-1, OR7-21
Chromosome Location
Chr. 1: 683910 - 720118 on Build GRCh38
UniGene ID
Hs.632360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017002408.1 XP_016857897.1 3 373
XM_017002409.1 XP_016857898.1 4 291
XM_017002410.1 XP_016857899.1 2 155
XM_017002411.1 XP_016857900.1 2 144

Gene Symbol
LOC101927506
Entrez Gene ID
101927506
Gene Name
uncharacterized LOC101927506
Gene Aliases
Chromosome Location
Chr. 8: 62551 - 208057 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017014132.1 XP_016869621.1 4 3994

Gene Symbol
OR4F3
Entrez Gene ID
26683
Gene Name
olfactory receptor family 4 subfamily F member 3
Gene Aliases
Chromosome Location
Chr. 5: 181342142 - 181370030 on Build GRCh38
UniGene ID
Hs.632360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017009338.1 XP_016864827.1 2 1856

View Full Product Details