Gene Transcripts

Gene Symbol
GPR17
Entrez Gene ID
2840
Gene Name
G protein-coupled receptor 17
Gene Aliases
Chromosome Location
Chr. 2: 127645864 - 127652639 on Build GRCh38
UniGene ID
Hs.46453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161415.1 NP_001154887.1 4 1671
NM_001161416.1 NP_001154888.1 3 1606
NM_001161417.1 NP_001154889.1 2 1377
NM_005291.2 NP_005282.1 3 1442
XM_017003833.1 XP_016859322.1 2 1798
GenBank mRNA AK126849.1 - 1 2632
AK292725.1 - 2 1247
AK313949.1 - 2 1133
BC031653.1 - 2 1056 5174146
BC039595.1 - 2 1061
JF810891.1 - 1 976
Y12546.1 - 3 1134
Z94154.1 - 2 1070
Z94155.1 - 1 854

View Full Product Details