Gene Transcripts
- Gene Symbol
- CDC26
- Entrez Gene ID
- 246184
- Gene Name
- cell division cycle 26
- Gene Aliases
- ANAPC12, APC12, C9orf17
- Chromosome Location
- Chr. 9: 113267008 - 113275589 on Build GRCh38
- UniGene ID
- Hs.727648
- Species
- Homo sapiens
- Gene Symbol
- FKBP15
- Entrez Gene ID
- 23307
- Gene Name
- FK506 binding protein 15
- Gene Aliases
- FKBP133, KIAA0674, PPP1R76
- Chromosome Location
- Chr. 9: 113165036 - 113221361 on Build GRCh38
- UniGene ID
- Hs.522351
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK307507.1 |
- |
3 |
271 |
|
View Full Product Details