Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001079512.3 | NP_001072980.1 | 4 | 578 | |
NM_001318873.1 | NP_001305802.1 | 4 | 1029 | ||
NR_134901.1 | 5 | 648 | |||
NR_134902.1 | 4 | 578 | |||
NR_134903.1 | 4 | 540 | |||
NR_134904.1 | 4 | 503 | |||
NR_134905.1 | 3 | 433 | |||
NR_134906.1 | 3 | 433 | |||
XM_006720944.3 | XP_006721007.1 | 4 | 686 | ||
XM_011522654.2 | XP_011520956.1 | 4 | 686 | ||
XM_017023649.1 | XP_016879138.1 | 4 | 686 | ||
XM_017023650.1 | XP_016879139.1 | 4 | 886 | ||
XM_017023651.1 | XP_016879140.1 | 4 | 314 | ||
GenBank mRNA | AK124106.1 | - | 4 | 886 | |
AK294418.1 | - | 4 | 569 | ||
AL833564.1 | - | 2 | 88 | ||
BC050569.1 | - | 4 | 353 | ||
BC146882.1 | - | 4 | 384 | 9056810 | |
BC146886.1 | - | 4 | 384 | 9056814 | |
BC171807.1 | - | 3 | 239 | ||
BC171816.1 | - | 3 | 239 | ||
BC171817.1 | - | 4 | 309 | 9054471 | |
BC171819.1 | - | 5 | 454 | ||
CN301583.1 | - | 4 | 548 |