Gene Transcripts

Gene Symbol
Aebp1
Entrez Gene ID
11568
Gene Name
AE binding protein 1
Gene Aliases
ACLP
Chromosome Location
Chr. 11: 5861853 - 5872225 on Build GRCm38
UniGene ID
Mm.3317
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF053943.1 - 21 3426
AK004712.1 - 17 2435
AK128980.1 - 21 3502
AK158276.1 - 21 3452
AK159330.1 - 21 3433
AK159342.1 - 21 3452
AK159377.1 - 21 3423
AK159409.1 - 21 3453
AK159774.1 - 21 3424
AK159957.1 - 21 3424
AK207752.1 - 1 129
AK216208.1 - 2 305
BC082577.1 - 21 3438 30648530
X80478.1 - 12 2156

View Full Product Details