Gene Transcripts

Gene Symbol
CXCL1
Entrez Gene ID
2919
Gene Name
C-X-C motif chemokine ligand 1
Gene Aliases
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1
Chromosome Location
Chr. 4: 73869392 - 73871302 on Build GRCh38
UniGene ID
Hs.708652
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001511.3 NP_001502.1 4 744
NR_046035.1 4 767
GenBank mRNA BC011976.1 - 4 732
BM554448.1 - 4 757
J03561.1 - 4 718
X12510.1 - 4 682

View Full Product Details