Gene Transcripts
- Gene Symbol
- Alk
- Entrez Gene ID
- 11682
- Gene Name
- anaplastic lymphoma kinase
- Gene Aliases
- CD246, Tcrz
- Chromosome Location
- Chr. 17: 71868988 - 72605410 on Build GRCm38
- UniGene ID
- Mm.311854
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007439.2 |
NP_031465.2
|
10 |
2484 |
|
View Full Product Details