Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001036684.2 | NP_001031761.1 | 12 | 2603 | |
NM_009723.5 | NP_033853.1 | 14 | 2792 | ||
GenBank mRNA | AB030737.1 | - | 11 | 2164 | |
AB030738.1 | - | 11 | 2164 | ||
AF053471.1 | - | 13 | 2621 | ||
AK054112.1 | - | 16 | 2890 | ||
AK147240.1 | - | 14 | 2678 | ||
AK147357.1 | - | 12 | 2532 | ||
AK147369.1 | - | 14 | 2727 | ||
AK147399.1 | - | 12 | 2755 | ||
AK147501.1 | - | 14 | 2739 | ||
GU734816.2 | - | 14 | 2299 |