Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001806.2 | NP_001001806.1 | 2 | 332 | |
GenBank mRNA | AK142565.1 | - | 138 | ||
AK159513.1 | - | 2 | 333 | ||
AK160955.1 | - | 2 | 331 | ||
AK170763.1 | - | 2 | 333 | ||
BB611978.1 | - | 2 | 331 | ||
BB656287.1 | - | 2 | 333 | ||
BC006189.1 | - | 1 | 181 | ||
BC028330.1 | - | 2 | 290 | ||
BC038863.1 | - | 2 | 54 | ||
BC139416.1 | - | 2 | 208 | 8862438 |