Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110504.1 | NP_001103974.1 | 10 | 1191 | |
NM_007600.3 | NP_031626.1 | 10 | 1243 | ||
GenBank mRNA | AF021847.1 | - | 10 | 1122 | |
AF084459.1 | - | 9 | 1109 | ||
AK088547.1 | - | 10 | 1163 | ||
AK149090.1 | - | 6 | 591 | ||
AK149706.1 | - | 10 | 1183 | ||
AK150084.1 | - | 10 | 1182 | ||
AK160940.1 | - | 10 | 1183 | ||
AK165258.1 | - | 10 | 1202 | ||
AK168063.1 | - | 10 | 1182 | ||
AK171405.1 | - | 10 | 1146 | ||
BC026138.1 | - | 10 | 1158 | 4218148 | |
BC050276.1 | - | 10 | 1159 | 4923347 |