Gene Transcripts
- Gene Symbol
- GYPB
- Entrez Gene ID
- 2994
- Gene Name
- glycophorin B (MNS blood group)
- Gene Aliases
- CD235b, GPB, GYP, MNS, PAS-3, SS
- Chromosome Location
- Chr. 4: 143995172 - 144021860 on Build GRCh38
- UniGene ID
- Hs.654368
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001304382.1 |
NP_001291311.1
|
4, 5 |
322 |
|
NM_002100.5 |
NP_002091.3
|
3, 4 |
242 |
|
XM_011531903.2 |
XP_011530205.1
|
3, 4 |
377 |
|
XM_011531904.2 |
XP_011530206.1
|
4, 5 |
2881 |
|
XM_011531905.2 |
XP_011530207.1
|
4, 5 |
529 |
|
XM_017008137.1 |
XP_016863626.1
|
3, 4 |
2576 |
|
GenBank mRNA |
AY950609.1 |
- |
3, 4 |
184 |
|
AY950610.1 |
- |
3, 4 |
184 |
|
AY950611.1 |
- |
3, 4 |
184 |
|
AY950612.1 |
- |
4, 5 |
277 |
|
BC069310.1 |
- |
3, 4 |
234 |
7262104 |
BC121077.1 |
- |
3, 4 |
218 |
40119713 |
BC121078.1 |
- |
3, 4 |
218 |
40119716 |
EU338223.1 |
- |
2, 3 |
122 |
|
EU338224.1 |
- |
4, 5 |
345 |
|
EU338225.1 |
- |
4, 5 |
314 |
|
EU338226.1 |
- |
4, 5 |
279 |
|
EU338228.1 |
- |
2, 3 |
122 |
|
EU338229.1 |
- |
4, 5 |
345 |
|
EU338235.1 |
- |
2, 3 |
122 |
|
EU338236.1 |
- |
4, 5 |
345 |
|
EU338237.1 |
- |
5, 6 |
438 |
|
EU338238.1 |
- |
1, 2 |
83 |
|
J02982.1 |
- |
3, 4 |
194 |
|
X08055.1 |
- |
5, 6 |
320 |
|
View Full Product Details