Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111060.2 | NP_001104530.1 | 5 | 502 | |
NM_007652.5 | NP_031678.1 | 4 | 382 | ||
GenBank mRNA | AK002743.1 | - | 4 | 331 | |
AK005507.1 | - | 4 | 350 | ||
AK018136.1 | - | 4 | 347 | ||
AK080728.1 | - | 4 | 330 | ||
BC132089.1 | - | 3 | 260 | 40130366 | |
BC132115.1 | - | 3 | 260 | 40130392 | |
CA467821.1 | - | 500 | |||
U60473.1 | - | 4 | 318 |