Gene Transcripts

Gene Symbol
Ch25h
Entrez Gene ID
12642
Gene Name
cholesterol 25-hydroxylase
Gene Aliases
AI462618, m25OH
Chromosome Location
Chr. 19: 34473783 - 34475135 on Build GRCm38
UniGene ID
Mm.30824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009890.1 NP_034020.1 1 608
GenBank mRNA AF059213.1 - 1 608
AK140360.1 - 1 626
AK152770.1 - 1 617
BC039919.1 - 1 641 4953185

View Full Product Details