Gene Transcripts

Gene Symbol
Ckmt1
Entrez Gene ID
12716
Gene Name
creatine kinase, mitochondrial 1, ubiquitous
Gene Aliases
Mt-CK, ScCKmit, UbCKmit, mi-CK
Chromosome Location
Chr. 2: 121357751 - 121363737 on Build GRCm38
UniGene ID
Mm.252145
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009897.2 NP_034027.1 5, 6 973
GenBank mRNA AK005076.1 - 5, 6 972
AK131953.1 - 5, 6 977
BC025976.1 - 6, 7 888 4216758
Z13968.1 - 5, 6 973

View Full Product Details