Gene Transcripts

Gene Symbol
Clk4
Entrez Gene ID
12750
Gene Name
CDC like kinase 4
Gene Aliases
AI987988, C85119
Chromosome Location
Chr. 11: 51263167 - 51281770 on Build GRCm38
UniGene ID
Mm.239354
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF005423.1 - 2 140
AF033566.1 - 5 569
AK161739.1 - 1 1578
AY653308.1 - 4 459
BC002220.1 - 1 125 3490616
BC012675.1 - 5 591 3982920
CT010293.1 - 4 491
U94846.1 - 5 588

View Full Product Details